missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemsplash Delta 67 Coverall with Knit Wrist Cuff Type 5 and 6 Hood

Click to view available options
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Unit Size:
Pack of 25
Description
Chemsplash Delta 67 Disposable Coverall
- 7GSM Microporous Laminate Fabric
- Elasticated 3 Piece Hood, Back and Ankles
- Low Linting Knitted Cuffs with Thumb Loops
- Two-Way Zip with Adhesive Zip Flap
- Reinforced Bound Seam at Crotch Area
- Non Linting and Anti-Static Fabric
- Silicone and Latex Free
- PFAS Free
- Conforms to ISO Cleanroom 14644-1:2015 Cleanroom Class 5-9
Description: The Chemsplash Delta 67 Disposable Coverall is made from soft, comfortable 67GSM microporous laminate fabric, meeting Cat III Type 5B/6B. It is compliant with EN1149-5:2008 (Antistatic) and EN1073-2 (Nuclear Particle). The coverall is ideal for various applications, particularly for prolonged wear, offering improved breathability while maintaining an effective barrier against hazards.
Specifications
Specifications
| Product Type | Cleanroom Coveralls |
| Size | Small |
| Gender | Unisex |
| Material | Microporous Composite Fabric |
| Color | White |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Knit, Thumb Loop |
| Neck Style | Hooded |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction