Learn More
Chemsplash Xtreme 50 SMS T5/6 Disposable Coverall Red

Description
Chemsplash Xtreme SMS 50 Coveralls
- 50GSM Breathable SMS Fabric
- Elasticated 3 Piece Hood
- Elasticated Cuffs and Ankles
- Two Way Zip with Adhesive Zip Flap
- Silicone and Latex Free
- Comes without the chest label
- PFAS Free
- See Style 2618 for White Sizes 4XL-5XL-6XL
Description: Chemsplash Xtreme SMS 50 disposable coveralls are made from extremely breathable SMS fabric suitable for prolonged periods of use. Featuring an elasticated hood, cuffs, and ankles, with a two-way zip and an adhesive cover flap. Its strong and robust properties provide a good chemical barrier, especially to dry hazardous particles, whilst being extremely comfortable to wear. Offering the best value within our Cat III Type 5 and 6 range. Available in White, Blue, Red, and Orange.
Specifications
Specifications
| Product Type | Hooded Coveralls |
| Size | Medium |
| Gender | Unisex |
| Material | SMS Fabric |
| Color | Red |
| Disposable | Single-use |
| Waist Style | Elastic |
| Ankle Style | Elastic |
| Cuff Style | Elastic |
| Neck Style | Hooded |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.