Product Code. 12656797
Shop All Fisher Chemical Products
Click to view available options
Packaging:
Aluminum Can
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
Quantity
Packaging
Unit Size
12656797
5 L
Aluminum Can
5 L
10355960
2.5 L
Amber Glass Bottle
2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code.
12656797
Supplier
Fisher Chemical
Supplier No.
X/0255/21
Estimated Shipment: 01-12-2026Log in to see stock.
missing translation for 'validMainframeMsg'
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC. We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Chemical Identifiers
CAS
1330-20-7
Molecular Formula
C8 H10
MDL Number
77264
Specifications
Melting Point
-34°C
Color
Colorless
Quantity
5 L
Packaging
Aluminum Can
Physical Form
Liquid
Vapor Pressure
8mbar at 20°C
Viscosity
0.6 mPaS at 20°C
Boiling Point
136°C
Formula Weight
106.17g/mol
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our
Privacy Policy .
Spot an opportunity for improvement?