missing translation for 'onlineSavingsMsg'
Learn More

Xylene, Extra Pure, SLR, Fisher Chemical™

Product Code. 11498922 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
5 L
25 L
Packaging:
Amber Glass Bottle
HDPE Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
5 L
500mL
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11498922 500 mL Amber Glass Bottle 500mL
10784001 2.5 L Amber Glass Bottle 2.5 L
10335942 2.5 L Plastic-coated Glass Bottle 2.5 L
10090400 5 L HDPE Bottle 5 L
10142260 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11498922 Supplier Fisher Chemical Supplier No. X/0200/08
Spotlight Offers
Use promo code 28036
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Xylene, Extra Pure, SLR

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
MDL Number 77264

Specifications

Melting Point -34°C
Color Colorless
Quantity 500 mL
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Boiling Point 136°C
Formula Weight 106.17g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.