missing translation for 'onlineSavingsMsg'
Learn More

Xylene, MP Biomedicals™

Product Code. 11449911 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
4 L
Unit Size:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11449911 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11449911 Supplier MP Biomedicals Supplier No. 0215869204
  • ONLINE EXCLUSIVE PRICE
    164.00 €
    155.40 € / 4 L
    Save 8.60 €
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Xylene, MP Biomedicals is a solvent used in Giemsa cell staining protocols.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1330-20-7
Molecular Formula C8H10
Synonym Xylol, dimethylbenzene, Xylene mixture of isomers
Solubility Information Soluble in absolute alcohol, ether, many organic solvents; practically in insoluble in water.
Formula Weight 106.17
Grade Histological
Quantity 4 L
Physical Form Liquid
compliance-icons
Product Identifier
  • 1-Bromo-3,5-bis(trifluoromethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1-Bromo-3,5-bis(trifluoromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.