missing translation for 'onlineSavingsMsg'
Learn More

Xylene, Technical, Fisher Chemical™

Product Code. 10588070 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
25 L
Packaging:
Amber Glass Bottle
Metal Can
Plastic-coated Glass Bottle
Unit Size:
2.5 L
25 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10588070 2.5 L Amber Glass Bottle 2.5 L
10691373 2.5 L Plastic-coated Glass Bottle 2.5 L
10305870 25 L Metal Can 25 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10588070 Supplier Fisher Chemical Supplier No. X/0100/17
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

Chemical Identifiers

CAS 1330-20-7
Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number 77264
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
IUPAC Name 1,3-xylene
SMILES CC1=CC(C)=CC=C1

Specifications

Melting Point -34°C
Color Colorless
Quantity 2.5 L
Packaging Amber Glass Bottle
Physical Form Liquid
Vapor Pressure 8mbar at 20°C
Viscosity 0.6 mPaS at 20°C
Boiling Point 136°C
Formula Weight 106.17g/mol
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Aspiration toxicity Category 1
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Aromatic hydrocarbons, C8; Light Oil Redistillate, high boiling, Xylenes (o-, m-, p- isomers), Ethyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.