missing translation for 'onlineSavingsMsg'
Learn More

Honeywell Xylenes

Product Code. 15935430
Change view
Click to view available options
Quantity:
45 L
Unit Size:
45 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15935430 45 L 45 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 15935430 Supplier Honeywell Supplier No. 1644645L

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Xylenes, Puriss., Assay xylene isomers and ethelbenzol (GC) ≥ 99%.

Supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:

  • Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
  • Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.

Specifications

pH Not applicable
Melting Point -34°C
CAS 1330-20-7
Color Colorless
CAS Max % ≤100.00%
Density 0.87 g/cm3
Quantity 45 L
Flash Point 25°C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid
Product Line Puriss.
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
Vapor Pressure 5 to 10 hPa (20°C)
MDL Number MFCD00077264
Viscosity 0.61 mPa.s (20°C)
UN Number UN1307
Synonym Xylene mixture of isomers
Boiling Point 136°C to 140°C
InChI Key IVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILES CC1=CC(C)=CC=C1
IUPAC Name 1,3-xylene
Molecular Weight (g/mol) 106.17
Percent Purity ≥99.00%
Grade Reagent
Chemical Name or Material Xylenes
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylenes
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
  • Aspiration toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312 + H332-Harmful in contact with skin or if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.