missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, 99+%, for residue analysis, mixed, electronic use grade

Product Code. 10771171 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10771171 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 10771171 Supplier Thermo Scientific Acros Supplier No. 327950010
  • ONLINE EXCLUSIVE PRICE
    87.90 €
    80.80 € / 1 L
    Save 7.10 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 1330-20-7 | C8H10

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point -34.0°C
CAS 1330-20-7, 100-41-4
Density 0.8650g/mL
Quantity 1 L
Flash Point 23°C
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 0.0005% max.
Assay Percent Range 99+%
Specific Gravity 0.865
Molecular Formula C8H10
Linear Formula C6H4(CH3)2
MDL Number MFCD00077264
Viscosity 0.6 mPa.s (20°C)
Water 0.02% max. (Coulometric)
Beilstein 05, 360
Merck Index 15, 10278
Synonym Dimethylbenzene
Boiling Point 136.0°C to 140.0°C
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity ≥99%
Grade Residue Analysis
Chemical Name or Material Xylenes, Residue Free
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Xylenes
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.