missing translation for 'onlineSavingsMsg'
Learn More

Xylenes, Semiconductor Grade

Product Code. 11353577
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 1 L
Unit Size:
1 L
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11353577 1 L 1 L
11343577 4 x 1 L 4 L
11363577 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 11353577 Supplier Thermo Scientific Alfa Aesar Supplier No. 019402.K2
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

CAS: 1330-20-7 | (CH3)2C6H4

Xylene is used as a solvent in pharmaceutical, printing, rubber, and leather industries. It is an important component of ink, rubber and adhesives as well as in thinning paints and varnishes. It is used in the laboratory to make baths with dry ice to cool reaction vessels, and as a solvent to remove synthetic immersion oil from the microscope objective in light microscopy. It finds application as a short-term hydrophobic clearing agent of tissue samples by histologists. It is a precursor for the preparation of xylene dichlorides, xylyl bromide, terephthalic acid and dimethyl terephthalate. It acts as a cleaning agent for steel, silicon wafers and integrated circuits.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1330-20-7
Molecular Formula (CH3)2C6H4
MDL Number MFCD00077264

Specifications

Melting Point <°C to 25°C
Color Colorless
Appearance Colorless liquid
Density 0.86
Quantity 1 L
Flash Point 29°C (84°F)
Ignition Point 465°C (869°F)
Odor Aromatic
Physical Form Liquid
Refractive Index 1.497
Vapor Pressure 10 hPa (8mm Hg) at 20°C
Beilstein 1901563
Boiling Point 137°C to 144°C
Solubility Information Miscible with aromatic hydrocarbons. Immiscible with water.
Formula Weight 106.17
Grade Semiconductor
Chemical Name or Material Xylenes
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.