missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange, BAKER™, J.T. Baker™

Product Code. 15578844 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Wide Mouth Amber Glass Bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15578844 5 g Wide Mouth Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15578844 Supplier Avantor J.T.Baker Supplier No. X58401

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A grade of chemicals for laboratory use

Chemical Identifiers

CAS 3618-43-7
Molecular Formula C31H28N2Na4O13S

Specifications

Formula Weight 760.6
Packaging Wide Mouth Amber Glass Bottle
Quantity 5 g
Chemical Name or Material Xylenol Orange
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.