missing translation for 'onlineSavingsMsg'
Learn More

Xylenol Orange Tetrasodium Salt, Indicator for Metal Titration, Solstice

Product Code. 15654090
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15654090 5 g Glass Bottle 5g
15644090 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15654090 Supplier Honeywell-Fluka Supplier No. 338255G
  • ONLINE EXCLUSIVE PRICE
    177.00 €
    148.95 € / 5g
    Save 28.05 €
    15% Off
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Indicator for metal titration

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3618-43-7
MDL Number MFCD00044293

Specifications

Melting Point 195°C
Packaging Glass Bottle
UN Number NONH for all modes of transport
Color Brown
Quantity 5 g
Physical Form Powder or Crystals
Molecular Weight (g/mol) 760.58
Formula Weight 760.58g/mol
Percent Purity 100%
Chemical Name or Material Xylenol Orange Tetrasodium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.