missing translation for 'onlineSavingsMsg'
Learn More

Xylidine Ponceau 2R, pure

Product Code. p-212340 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10012450

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Xylidine Ponceau, CAS # 3761-53-3, is a red azo dye used in histology for staining of thin sections. | CAS: 3761-53-3 | C18H14N2Na2O7S2 | 480.417 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Xylidine Ponceau 2R is largely confined to Masson's trichrome, where it gives a slight orange shading to the red of the cytoplasmic structures

Application

  • Xylidine Ponceau 2R is a histological dye used mainly for the staining of thin sections
  • It shows cardiotoxic and carcinogenic activities
  • It has been used to prepare Fuchsin-Ponceau solution to stain an amyotrophic lateral sclerosis tissue-engineered skin model
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3761-53-3
Molecular Formula C18H14N2Na2O7S2
Molecular Weight (g/mol) 480.417
MDL Number MFCD00003897
InChI Key YJVBLROMQZEFPA-UHFFFAOYSA-L
Synonym 4-(2,4-Dimethylphenylazo)-3-hydroxy-2,7-naphthalenedisulfonic acid, disodium salt, C.I. 16150, 4-(2, 4-Dimethylphenylazo)-3-hydroxy-2, 7-naphthalenedisulfonic acid, Ponceau G, R, 2R
PubChem CID 102092631
ChEBI CHEBI:82369
IUPAC Name disodium;5-[(2,4-dimethylphenyl)diazenyl]-6-oxido-7-sulfonaphthalene-2-sulfonate
SMILES CC1=CC(=C(C=C1)N=NC2=C3C=CC(=CC3=CC(=C2[O-])S(=O)(=O)O)S(=O)(=O)[O-])C.[Na+].[Na+]

Specifications

Infrared Spectrum Authentic
Formula Weight 480.41
Grade Extra Pure
Packaging Glass bottle
UV Spectrometry Complies
Color Bordeaux to Red
Quantity 25 g
Physical Form Fine Powder
Chemical Name or Material Xylidine Ponceau 2R
compliance-icons compliance-icons
Product Identifier
  • Xylidine Ponceau 2R
Signal Word
  • Warning
Hazard Category
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.