missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Xylitol, 99%

Product Code. 11418786
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11418786 500 g 500g
11408786 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11418786 Supplier Thermo Scientific Chemicals Supplier No. A16944.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as a polyol substrate for xylitol and sorbitol dehydrogenases and sorbitol dehydrogenase derived from xylose. It is applied as a sweetener and humectants and skin conditioning agent in cosmetics industry.

Solubility
Soluble in water (50 mg/ml), abs methanol (6 g/100ml), abs ethanol (1.2 g/100ml), alcohol (very slightly), and pyridine. Insoluble in ether.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Store under inert gas.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92°C to 96°C
Density 1.515
Assay Percent Range 99%
Odor Odorless
Quantity 500 g
Beilstein 1720523
Sensitivity Hygroscopic
Merck Index 14,10085
Solubility Information Soluble in water (50mg/ml),abs methanol (6g/100ml),abs ethanol (1.2g/100ml),alcohol (very slightly),and pyridine. Insoluble in ether.
IUPAC Name pentane-1,2,3,4,5-pentol
Formula Weight 152.15
Percent Purity 99%
Chemical Name or Material Xylitol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.