missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, 99+%

Product Code. 10276610 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
25g
500g
This item is not returnable. View return policy

Product Code. 10276610

Brand: Thermo Scientific Acros 225981000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 87-99-0 | C5H12O5 | 152.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87-99-0
Molecular Formula C5H12O5
Molecular Weight (g/mol) 152.15
MDL Number MFCD00064291,MFCD00064292
InChI Key HEBKCHPVOIAQTA-UHFFFAOYNA-N
Synonym xylitol, ribitol, adonitol, d-xylitol, xylite, adonit, adonite, xylit, d-ribitol, eutrit
PubChem CID 6912
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
SMILES OCC(O)C(O)C(O)CO

Specifications

Melting Point 92.0°C to 96.0°C
Color White
Boiling Point 215.0°C to 217.0°C
Infrared Spectrum Authentic
Assay Percent Range 2% max. Other polyols (GC)
Packaging Plastic bottle
Linear Formula HOCH2[CH(OH)]3CH2OH
Quantity 100 g
Beilstein 01, 531
Merck Index 15, 10283
Solubility Information Solubility in water: 1700g/L (20°C). Other solubilities: soluble in pyridine,less soluble in alcohols
Formula Weight 152.15
Percent Purity 99+%
Physical Form Crystalline Powder
Chemical Name or Material Xylitol, 99+%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.