missing translation for 'onlineSavingsMsg'
Learn More

Xylitol, TRC

Product Code. 30457354
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30457354 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30457354 Supplier Toronto Research Chemicals Supplier No. TRCX748050500G
 Restricted Item
Estimated Shipment: 18-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Xylitol
CAS 87-99-0
Molecular Formula C5 H12 O5
Quantity 500 g
Synonym Xylitol','Meso-xylitol','C-Xylidex CR 16055','DL-Xylitol','Eutrit','Fluorette','Klinit','Kylit','NSC 25283','Wood sugar alcohol','X 0018','Xylisorb','Xylisorb 300','Xylisorb 700','Xylit','Xylit XC','Xylitab 100','Xylitab 300','Xylitab DC','Xylite','Xylite (sugar)','Xylitol C','Xylitol CM 90','Xylitol P','Xyliton','meso-Xylitol','xylo-Pentitol
InChI Formula InChI=1S/C5H12O5/c6-1-3(8)5(10)4(9)2-7/h3-10H,1-2H2/t3-,4+,5+
SMILES OC[C@@H](O)[C@H](O)[C@@H](O)CO
IUPAC Name (2S,4R)-pentane-1,2,3,4,5-pentol
Molecular Weight (g/mol) 152.15
Formula Weight 152.0685
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.