missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Y-27632 dihydrochloride

Product Code. 30021997
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30021997 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30021997 Supplier R&D Systems™ Supplier No. TB1254GMP/10
  • ONLINE EXCLUSIVE PRICE
    1636.97 €
    1452.15 € / 10mg
    Save 184.82 €
    11% Off
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Y-27632 synthesized to cGMP guidelines

Y-27632 dihydrochloride is a Y-27632 synthesized to cGMP guidelines. Suitable for use as an ancillary reagent in the further manufacturing of cell therapies.

For more information about how Y-27632 may be used, please see our protocols: Transdifferentiating Fibroblasts into Neurons, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts .

Research use only Y-27632 also available.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide dihydrochloride
CAS 129830-38-2
Quantity 10 mg
Product Line Tocris
Target Rho-kinase Inhibitors
Molecular Formula C14H21N3O.2HCl
Purity 99%
Regulatory Status GMP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.