missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies Y-27632 (Dihydrochloride)

Product Code. 15427440
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
50 mg
5 x 10 mg
Unit Size:
1mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15427440 1 mg 1mg
15537014 50 mg 50mg
15580821 5 mg 5mg
15627226 5 x 10 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15427440 Supplier STEMCELL Technologies Supplier No. 72302
 Restricted Item
Estimated Shipment: 07-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

RHO/ROCK pathway inhibitor; Inhibits ROCK1 and ROCK2

  • Y-27632 is a cell-permeable, highly potent and selective inhibitor of Rho-associated, coiled-coil containing protein kinase (ROCK)
  • Y-27632 inhibits both ROCK1 (Ki = 220 nM) and ROCK2 (Ki = 300 nM) by competing with ATP for binding to the catalytic site

Specifications

CAS 129830-38-2
Quantity 1 mg
Molecular Formula C14H21N3O · 2HCl
Purity ≥98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.