missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Y-27632 (Dihydrochloride)
Click to view available options
Quantity:
1 mg
5 mg
50 mg
5 x 10 mg
Unit Size:
1mg
50mg
5mg
Description
- Y-27632 is a cell-permeable, highly potent and selective inhibitor of Rho-associated, coiled-coil containing protein kinase (ROCK)
- Y-27632 inhibits both ROCK1 (Ki = 220 nM) and ROCK2 (Ki = 300 nM) by competing with ATP for binding to the catalytic site
Specifications
Specifications
| CAS | 129830-38-2 |
| Quantity | 5 x 10 mg |
| Molecular Formula | C14H21N3O · 2HCl |
Safety and Handling
Product Identifier
- Y-27632 (Dihydrochloride)
- Warning
- Acute toxicity Category 4
- H302-Harmful if swallowed.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
- MIXTURE LIST-Contains : 4-[(1R)-1-aminoethyl]-N-pyridin-4-ylcyclohexane-1-carboxamide;dihydrochloride
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto