missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Y-27632 dihydrochloride

Product Code. 18084362
Change view
Click to view available options
Quantity:
1 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18084362 10 mg 10mg
30001217 1 mg 1mg
18044982 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 18084362 Supplier Tocris Bioscience™ Supplier No. 1254/10
  • ONLINE EXCLUSIVE PRICE
    435.00 €
    410.55 € / 10mg
    Save 24.45 €
    5% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Selective p160ROCK inhibitor

Y-27632 dihydrochloride is a selective ROCK inhibitor (Ki values are 0.14-0.22, 0.3, 25, 26 and > 250 μM for ROCK1 (p160 ROCK), ROCK2, PKA, PKC and MLCK respectively). Also inhibits PRK2 (IC50 = 600 nM). Increases survival rate of human embryonic stem (hES) cells and iPSC undergoing cryopreservation. Used in combination with CHIR99021, RepSox, Forskolin, SP 600125, Go 6983 and VPA to reprogram fibroblasts to mature neurons. Also optimizes naive human pluripotent stem cell growth and viability following naive cell derivation from primed ESCs and iPSCs using naive human stem cell medium (NHSM). Used as component of growth media for urothelial organoids. Cell-permeable, orally bioavailable and active in vivo. Y-27632 synthesized to cGMP guidelines also available.

For more information about how Y-27632 dihydrochloride may be used, see our protocols: Transdifferentiating Fibroblasts into Neurons, Converting Fibroblasts into Cardiomyocytes (9C Cocktail), Accelerated Induction of Cortical Neurons from hiPSCs, Human iPSC-derived Myoblasts .

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129830-38-2
Molecular Formula C14H23Cl2N3O

Specifications

Quantity 10 mg
Formula Weight 320.26
Purity >98%
Molecular Weight (g/mol) 320.258
Chemical Name or Material Y-27632 dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.