missing translation for 'onlineSavingsMsg'
Learn More

Y-27632 Dihydrochloride, TRC

Product Code. 30010890
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010890 10 mg 10mg
30008525 50 mg 50mg
30009040 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010890 Supplier Toronto Research Chemicals Supplier No. TRCD45544310MG
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 129830-38-2
Molecular Formula C14H21N3O • 2(HCl)
Molecular Weight (g/mol) 247.3423646
Synonym trans-4-[(1R)-1-Aminoethyl]-N-4-pyridinylcyclohexanecarboxamide Dihydrochloride, (+)-(R)-trans-4-(1-aminoethyl)-N-(4-pyridyl)cyclohexanecarboxamide Dihydrochloride, (+)-trans-N-(4-Pyridyl)-4-[(R)-1-aminoethyl]cyclohexanecarboxamide Dihydrochloride;
SMILES N[C@@H]([C@]1(CC[C@@H](CC1)C(NC2=CC=NC=C2)=O)[H])C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C14H21N3O.2ClH/c1-10(15)11-2-4-12(5-3-11)14(18)17-13-6-8-16-9-7-13;;/h6-12H,2-5,15H2,1H3,(H,16,17,18);2*1H/t10-,11?,12?;;/m1../s1
Formula Weight 319.1218
Percent Purity >95%
Chemical Name or Material Y-27632 Dihydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.