missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Y-27632 in solution

Product Code. 30067463
Change view
Click to view available options
Quantity:
4 mg
Unit Size:
4mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30067463 4 mg 4mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30067463 Supplier R&D Systems™ Supplier No. 7000/4
  • ONLINE EXCLUSIVE PRICE
    378.55 €
    339.15 € / 4mg
    Save 39.40 €
    10% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Sterile-filtered 10mM solution of Y-27632 pre-dissolved in water

Y-27632 in solution is a convenient and ready-to-use 10 mM sterile-filtered solution of Y-27632 (Cat. No. 1254), pre-dissolved in water. Increases survival rate of human embryonic stem (hES) cells and iPSC undergoing cryopreservation. Optimizes naive human pluripotent stem cell growth and viability following naive cell derivation from primed ESCs and iPSCs using naive human stem cell medium (NHSM). Used as a component of growth media for urothelial organoids and in a protocol to generate brain organoids from human iPSCs.

Supplied in vials containing a minimum of 1 mL of Y-27632 solution.

TRUSTED_SUSTAINABILITY

Spécification

CAS 129830-38-2
Quantity 4 mg
Product Line Tocris
Target Rho-kinase Inhibitors
Molecular Formula C14H21N3O.2HCl
Purity 0.97
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.