missing translation for 'onlineSavingsMsg'
Learn More

Y5, Thermo Scientific Chemicals

Product Code. 17176544
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
Unit Size:
100mg
250mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17176544 250 mg 250mg
17166544 100 mg 100mg
17186544 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17176544 Supplier Thermo Scientific Alfa Aesar Supplier No. H66460.MD
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2304444-48-0 | C{8}{2}H{9}{0}N{8}O{2}S{5} | 1379.97 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2304444-48-0
Molecular Formula C{8}{2}H{9}{0}N{8}O{2}S{5}
Synonym (2,2’-((2 Z,2’Z)-((12,13-bis(2-ethylhexyl)-3,9-diundecyl-12,13-dihydro[1,2,5 ]thiadiazolo[3,4 e]thieno[2’’,3’’:4’,5’] thieno[2’,3’:4,5 ]pyrrolo[3,2-g] thieno[2’,3’:4,5 ]thieno[3,2-b]indole-2,10-diyl)bis(methanylylidene))bis(3-oxo-2,3-dihydro1H-indene-2,1-diylidene))dimalononitrile)

Specifications

Quantity 250 mg
Formula Weight 1379.97
Physical Form Solid
Chemical Name or Material Y5

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.