missing translation for 'onlineSavingsMsg'
Learn More

Yeast Extract, Powder, MP Biomedicals™

Product Code. 11486501 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11486501 250 g 250g
11496501 1 kg 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486501 Supplier MP Biomedicals Supplier No. 0219402783

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Yeast extract From Yeast, Molecular Biology Reagent, Yellow-tan Powder

  • It is used as a bacteriological media component for a variety of microorganisms and molecular genetics applications.
  • Yeast extract powder is a vacuum dried autolyzed yeast extract produced from 100% Baker's yeast.
  • It is a concentrated source of soluble protein components, carbohydrates, and B Complex vitamins natural to yeast.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8013-01-2

Specifications

Color Tan-Yellow
Form Solid
Purity or Quality Grade Molecular Biology Grade
Quantity 250 g
Chemical Name or Material Yeast Extract
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.