missing translation for 'onlineSavingsMsg'
Learn More

Kartell Yellow Polypropylene Box, without Tips, with Standard Cover

Product Code. 18097761
Change view
Click to view available options
Color:
Yellow
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
18097761 Yellow Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18097761 Supplier Kartell Supplier No. 0975606
  • ONLINE EXCLUSIVE PRICE
    38.00 €
    36.10 € / Pack of 10
    Save 1.90 €
    5% Off
Estimated Shipment: 23-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Empty yellow PP boxes with snap-on covers for use with refill tips with capacity up to 1000 μL.

  • Empty box
  • For racked pipette tips

Specifications

Autoclavable Not Autoclavable
Color Yellow
Holds Pipette Tips
Material Polypropylene
Shape Rectangular
Stackable No
Sterility Non-sterile
For Use With (Equipment) Tips up to 1000 μL
Length (Metric) 11.50 cm
Product Type Empty Tip Box
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.