missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ YM 254890

Product Code. 18739646
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18739646 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18739646 Supplier R&D Systems™ Supplier No. 7352/1
  • ONLINE EXCLUSIVE PRICE
    625.12 €
    560.70 € / 1mg
    Save 64.42 €
    10% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Gα-protein inhibitor; potent inhibitor of Gq/11 signaling

YM 254890 is a Gα-protein inhibitor. Potently inhibits Gq/11 heterotrimeric G protein signaling. Inhibits ATP/UTP-induced Ca2+ increase in HCAE cells expressing P2Y2 receptor (IC50 = 50 nM); carbachol-induced production of IP1 in CHO cells expressing M1 receptor (IC50 = 95 nM); and ADP-induced Ca2+ increase in platelets, in vitro (IC50 = 2 μM). Also inhibits Gs-induced cAMP elevation and G protein-coupled MAPK/ERK signaling.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material (R)-1-((3 S,6 S,9 S,12 S,18 R,21 S,22 R)-21-acetamido-18-benzyl-3-((R)-1-methoxyethyl)-4,9,10,12,16,22-hexamethyl-15-methylene-2,5,8,11,14,17,20-heptaoxo-1,19-dioxa-4,7,10,13,16-pentaazacyclodocosan-6-yl)-2-methylpropyl (2 S,3 R)-2-acetamido-3-hydroxy-4-methylpent
CAS 568580-02-9
Quantity 1 mg
Product Line Tocris
Target GTPase Inhibitors
Molecular Formula C46H69N7O15
Purity 0.95
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.