missing translation for 'onlineSavingsMsg'
Learn More

YM155, TRC

Product Code. 30452131
Change view
Click to view available options
Quantity:
10 mg
25 mg
Unit Size:
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30452131 25 mg 25mg
30449320 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30452131 Supplier Toronto Research Chemicals Supplier No. TRCY10030025MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 781661-94-7
Molecular Formula C20H19BrN4O3
Molecular Weight (g/mol) 443.29
Synonym Sepantronium bromide, YM 155, YM155
IUPAC Name 1-(2-methoxyethyl)-2-methyl-3-(pyrazin-2-ylmethyl)benzo[f]benzimidazol-3-ium-4,9-dione;bromide
SMILES [Br-].COCCn1c(C)[n+](Cc2cnccn2)c3C(=O)c4ccccc4C(=O)c13

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C20H19N4O3.BrH/c1-13-23(9-10-27-2)17-18(24(13)12-14-11-21-7-8-22-14)20(26)16-6-4-3-5-15(16)19(17)25;/h3-8,11H,9-10,12H2,1-2H3;1H/q+1;/p-1
Formula Weight 442.0641
Chemical Name or Material 4,9-Dihydro-1-(2-methoxyethyl)-2-methyl-4,9-dioxo-3-(2-pyrazinylmethyl)-1H-naphth[2,3-d]imidazolium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.