missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yohimbine hydrochloride, 98+%

Product Code. 15432999
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15432999 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15432999 Supplier Thermo Scientific Chemicals Supplier No. J60185.03
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 65-19-0 | C21H27ClN2O3, C21H26N2O3·HCl | 390.91 g/mol

Yohimbine hydrochloride is an α2-adrenergic receptor antagonist. It blocks the central α2-adrenegic receptors in the brain, thus preventing and reducing the effects of xylazine, an α2-adrenergic agonist. The sedative effects and respiratory depression that accompany xylazine administration are both antagonized and reversed by yohimbine. Yohimbine hydrochloride also produces an antidiuretic effect, and increases heart rate and blood pressure.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 g
Description White to Off-white
Format Powder
For Use With (Application) Standard alpha-2-adrenergic antagonist
CAS 65-19-0
Molecular Formula C21H26N2O3·HCl
Formula Weight 390.91g/mol
Synonym 17-Hydroxyyohimban-16-carboxylic acid methyl ester hydrochloride
Sensitivity Light Sensitive
Melting Point 288°C to 290°C
Product Type Signal Transduction Reagent
Form Powder
Assay ≥98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.