missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Yohimbine hydrochloride, 99%

Product Code. 10356450 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10356450

Brand: Thermo Scientific Chemicals undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Yohimbine hydrochloride, CAS # 65-19-0, is an α2-adrenergic receptor antagonist, which blocks the central α2-adrenegic receptors in the brain. | CAS: 65-19-0 | C21H27ClN2O3 | 390.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Yohimbine hydrochloride is an indole alkaloid derived from the tree bark
  • It is an antagonist of α2-adrenergic receptors in the brain, reducing the effects of xylazine, which is an α2-adrenergic agonist

Applications

  • Yohimbine hydrochloride blocks the central α2-adrenegic receptors in the brain
  • It can reduce or eradicate effects of xylazine, which is an α2-adrenergic agonist
  • It can have an antidiuretic effect
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-19-0
Molecular Formula C21H27ClN2O3
Molecular Weight (g/mol) 390.91
MDL Number MFCD00012674
InChI Key PIPZGJSEDRMUAW-VJDCAHTMSA-N
Synonym yohimbine hydrochloride, antagonil, yohimbe, yohimbine hcl, aphrodine hydrochloride, yohimbine monohydrochloride, unii-nb2e1yp49f, yohimbin hydrochloride usp, nb2e1yp49f
PubChem CID 6169
IUPAC Name methyl (1S,15R,18S,19R,20S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
SMILES [H+].[Cl-].COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCC4=C(NC5=CC=CC=C45)[C@@H]3C[C@H]12

Specifications

Melting Point 285°C to 288°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 10300
Specific Rotation Condition +99° (20°C c=1%,H2O,dry substance)
Specific Rotation + 99
Formula Weight 390.9
Percent Purity 99%
Physical Form Powder
Chemical Name or Material Yohimbine hydrochloride
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Yohimbine hydrochloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H373-May cause damage to organs.
  • H331-Toxic if inhaled.
  • H311-Toxic in contact with skin.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.