missing translation for 'onlineSavingsMsg'
Learn More

Yohimbine Hydrochloride, TRC

Product Code. 30010595
Change view
Click to view available options
Quantity:
50 mg
1 g
5 g
Unit Size:
1g
50mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30010595 5 g 5g
30005682 1 g 1g
30011824 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30010595 Supplier Toronto Research Chemicals Supplier No. TRCY1008005G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 65-19-0
Molecular Formula C21 H26 N2 O3 . Cl H
Molecular Weight (g/mol) 390.9
Synonym Yohimban-16-carboxylic acid, 17-hydroxy-, methyl ester, monohydrochloride, (16α,17α)- (9CI), Yohimban-16α-carboxylic acid, 17α-hydroxy-, methyl ester, monohydrochloride (8CI), Benz[g]indolo[2,3-a]quinolizine, yohimban-16-carboxylic acid deriv., Actibine, Antagonil, Aphrodyne, Dayto-Himbin, Erex, NIH 9689, NSC 19509, Thybine, Yocon, Yohimbe, Yohimbex, Yohimbine chloride, Yohimbine hydrochloride, Yohimbine monohydrochloride, Yohimex, Yomax
IUPAC Name methyl (1S,15R,18S,19R,20S)-18-hydroxy-1,3,11,12,14,15,16,17,18,19,20,21-dodecahydroyohimban-19-carboxylate;hydrochloride
SMILES Cl.COC(=O)[C@H]1[C@@H](O)CC[C@H]2CN3CCc4c([nH]c5ccccc45)[C@@H]3C[C@H]12

Specifications

Quantity 5 g
InChI Formula InChI=1S/C21H26N2O3.ClH/c1-26-21(25)19-15-10-17-20-14(13-4-2-3-5-16(13)22-20)8-9-23(17)11-12(15)6-7-18(19)24;/h2-5,12,15,17-19,22,24H,6-11H2,1H3;1H/t12-,15-,17-,18-,19+;/m0./s1
Formula Weight 390.171
Percent Purity >95%
Chemical Name or Material Yohimbine hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.