missing translation for 'onlineSavingsMsg'
Learn More

Melford YPD Broth

Product Code. p-200087125
Click to view available options
Quantity:
250 g
1 kg
5 kg
Unit Size:
1kg
250g
5kg
This item is not returnable. View return policy

Product Code. 30018957

Brand: Melford Y20090250.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For maintaining and propagating yeasts in molecular biology applications.

For maintaining and propagating yeasts in molecular biology applications. The addition of protein and yeast extract promotes better growth.

Dissolve 50 grams in 1 liter of distilled water. Sterilize by autoclaving for 15 minutes.

Ingredients:

Peptone: 20 g/l

Dextrose: 20 g/l

Yeast extract: 10 g/l

Specifications

Quantity 250 g
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.