Learn More
YSI™ Ammonium Calibration Solutions
Product Code. 12796945
Click to view available options
Description:
Concentration: 100mg/L
Concentration: 10mg/L
Concentration: 1mg/L
Unit Size:
Each
Product Code. 12796945
Brand: YSI™ 003842
Only null left
Add to basket
Add to basket
Extremely accurate calibration solutions to calibrate field instruments equipped with ammonium ISE sensors
  • Concentration: 10mg/mL
  • Packaging: Plastic bottle
Order Info
Dated Material. Item is heat packed in the winter months.
TRUSTED_SUSTAINABILITY
Specifications
Description Concentration: 10mg/L
Type Ammonium Calibration Solution
Quantity 500 mL