missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Ammonium Calibration Solutions
Click to view available options
Description:
Concentration: 100mg/L
Concentration: 10mg/L
Concentration: 1mg/L
Unit Size:
Each
Description
- Concentration: 100mg/mL
- Packaging: Plastic bottle
Order Info
Dated Material. Item is heat packed in the winter months.
Specifications
Specifications
| Description | Concentration: 100mg/L |
| Type | Ammonium Calibration Solution |
| Quantity | 500 mL |
| For Use With (Equipment) | Calibrating field ammonium ISE sensors |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction