missing translation for 'onlineSavingsMsg'
Learn More

YSI™ Buffer Solutions for pH Electrodes

Product Code. 12963347
Change view
Click to view available options
Color:
Blue
Yellow
pH Range:
7
10
Unit Size:
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color pH Range unitSize
12963347 Blue 10 Pack of 6
12953347 Yellow 7 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12963347 Supplier YSI™ Supplier No. 003823

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Designed and manufactured to the highest quality standards

  • Calibration buffer
  • For use with YSI 60, YSI 556, Ecosense™ pH10, Ecosense pH100, 6-Series sonde meters
  • Color coded: pH 4 (red), pH 7 (yellow), pH 10 (blue)

TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Type Calibration Buffer
pH Range 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.