missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YSI™ Buffer Solutions for pH Electrodes
Click to view available options
Color:
Blue
Yellow
pH Range:
7
10
Unit Size:
Pack of 6
Description
- Calibration buffer
- For use with YSI 60, YSI 556, Ecosense™ pH10, Ecosense pH100, 6-Series sonde meters
- Color coded: pH 4 (red), pH 7 (yellow), pH 10 (blue)
Specifications
Specifications
| Color | Yellow |
| Type | Calibration Buffer |
| pH Range | 7 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction