Learn More
YSI™ Membrane kits
Product Code. 12774516
Change view
Click to view available options
Probe Type:
DO
Product Code. Probe Type
12774516 DO
1 options
Product Code. 12774516 Supplier YSI™ Supplier No. 605306
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Specifications
For Use With (Equipment) YSI™ Model 556
Type Membrane Kit
Includes Six 1.25 mil. cap membranes, KCl solution
Probe Type DO