missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) chloride hexahydrate, 99.9%, (trace metal basis)

Product Code. 10637314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10637314 10 g Glass bottle 10g
10226300 1 g Glass bottle 1g
10607504 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10637314 Supplier Thermo Scientific Acros Supplier No. 194430100
 Restricted Item
Estimated Shipment: 16-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 10035-01-5 | Cl3Yb·6H2O | 387.48 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10035-01-5
Molecular Formula Cl3Yb·6H2O
Molecular Weight (g/mol) 387.48
MDL Number MFCD00149885

Specifications

Melting Point 865.0°C
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.90%
Linear Formula YbCl3·6H2O
Fieser 16,384
Merck Index 14, 10105
Solubility Information Solubility in water: soluble. Other solubilities: soluble in absolute alcohol
Formula Weight 387.48
Percent Purity 99.9%
Grade Trace Metal Basis
Packaging Glass bottle
Chemical Name or Material Ytterbium(III) chloride hexahydrate
Show More Show Less
compliance-icons
Product Identifier
  • Ytterbium(III) chloride hexahydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.