missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) oxide, 99.99%, (trace metal basis)

Product Code. 11631399 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11631399 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11631399 Supplier Thermo Scientific Supplier No. 264300100
 Restricted Item
Estimated Shipment: 07-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 1314-37-0 | O3Yb2 | 394.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 2346.0°C
CAS 1314-37-0
Color White
Physical Form Powder
Quantity 10 g
Assay Percent Range 99.99%
Molecular Formula O3Yb2
Linear Formula Yb2O3
MDL Number MFCD00011290
Merck Index 14,10105
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in hot dilute acids
InChI Key UZLYXNNZYFBAQO-UHFFFAOYSA-N
SMILES [O--].[O--].[O--].[Yb+3].[Yb+3]
IUPAC Name diytterbium(3+) trioxidandiide
Molecular Weight (g/mol) 394.09
Formula Weight 394.08
Percent Purity 99.99%
Grade Trace Metal Basis
Packaging Glass bottle
Density 9.17
Chemical Name or Material Ytterbium(III) oxide, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.