missing translation for 'onlineSavingsMsg'
Learn More

Ytterbium(III) Trifluoromethanesulfonate, TRC

Product Code. 30451125
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30451125 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30451125

Brand: Toronto Research Chemicals TRCY1200101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ytterbium(III) Trifluoromethanesulfonate
CAS 54761-04-5
Molecular Formula 3 C H F3 O3 S . Yb
Quantity 1 g
Synonym Trifluoromethanesulfonic Acid Ytterbium(3+) Salt, Tris(trifluoromethanesulfonato)ytterbium, Ytterbium Triflate, Ytterbium TrifluoromethanesulfonateYtterbium Tris(trifluoromethanesulfonate), Ytterbium Tritriflate, Ytterbium(3+) Triflate, Ytterbium(3+) Trifluoromethanesulfonate, Ytterbium(III) Triflate, Ytterbium(III) Trifluoromethanesulfonate
InChI Formula InChI=1S/3CHF3O3S.Yb/c3*2-1(3,4)8(5,6)7;/h3*(H,5,6,7);
SMILES [Yb].OS(=O)(=O)C(F)(F)F.OS(=O)(=O)C(F)(F)F.OS(=O)(=O)C(F)(F)F
IUPAC Name trifluoromethanesulfonic acid;ytterbium
Molecular Weight (g/mol) 623.271
Formula Weight 623.818
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.