missing translation for 'onlineSavingsMsg'
Learn More

Yttrium acetate hydrate, 99.9%, (trace metal basis)

Product Code. 10580574 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10580574 100 g Glass bottle 100g
10035123 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10580574 Supplier Thermo Scientific Acros Supplier No. 392211000
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 207801-28-3 | C6H9O6Y | 266.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207801-28-3
Molecular Formula C6H9O6Y
Molecular Weight (g/mol) 266.04
MDL Number MFCD00150128
InChI Key JUWHRRAPBUAYTA-UHFFFAOYSA-K
Synonym yttriumacetatetetrahydrate, yttrium acetate-water 1/3/4, yttrium 3+ triacetate tetrahydrate, yttrium 3+ ion triacetate tetrahydrate
PubChem CID 19818017
SMILES [Y+3].CC([O-])=O.CC([O-])=O.CC([O-])=O

Specifications

Color White
Physical Form Crystalline Powder and Lumps
Quantity 100 g
Assay Percent Range 99.90%
Solubility Information Solubility in water: soluble
IUPAC Name yttrium(3+);triacetate;tetrahydrate
Formula Weight 266.04
Percent Purity 99.9%
Grade Trace Metal Basis
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Yttrium acetate hydrate
Show More Show Less
compliance-icons
Product Identifier
  • Yttrium acetate hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.