missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Yttrium fluoride, 99.99%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Plastic bottle
Unit Size:
5g
Description
For ultra trace analysis
Specifications
Specifications
| Quantity | 5 g |
| Assay Percent Range | 99.99% |
| Formula Weight | 145.9 |
| Grade | Trace Metal Basis |
| Packaging | Plastic bottle |
| Chemical Name or Material | Yttrium fluoride |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction