missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) fluoride, anhydrous, 99.9% (metals basis)

Product Code. 11364128
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11364128

Brand: Thermo Scientific Alfa Aesar 041424.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For vacuum deposition | CAS: 13709-49-4 | F3Y | 145.90 g/mol

Yttrium(III) fluoride is used for the production of metallic yttrium, thin films, glasses, electronics and ceramics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13709-49-4
Molecular Formula F3Y
Molecular Weight (g/mol) 145.90
MDL Number MFCD00049714
InChI Key RBORBHYCVONNJH-UHFFFAOYSA-K
IUPAC Name yttrium(3+) trifluoride
SMILES [F-].[F-].[F-].[Y+3]

Specifications

Melting Point 1387°C
Physical Form Pieces
Quantity 50 g
UN Number UN3288
Solubility Information Soluble in acid. Insoluble in water.
Formula Weight 145.9
Percent Purity 99.9%
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Yttrium(III) fluoride, Anhydrous

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.