missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, 99.99%, (trace metal basis)

Product Code. 10199222 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Unit Size:
10g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10199222 10 g Glass bottle 10g
10193920 50 g Glass bottle 50g
15720829 250 g Glass bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10199222 Supplier Thermo Scientific Acros Supplier No. 194610100
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

for ultra trace analysis
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Specifications

Melting Point 2410.0°C
Color White
Physical Form Crystalline Powder
Quantity 10 g
Assay Percent Range 99.99% min. (Y2O3) TREO base
Linear Formula Y2O3
Merck Index 14, 10107
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in dilute acids
Formula Weight 225.81
Percent Purity 99.99%
Grade Trace Metal Basis
Loss on Ignition 1.0 % max. (1000°C) (material base)
Packaging Glass bottle
Chemical Name or Material Yttrium(III) oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.