missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) oxide, REacton™, 99.99% (REO)

Product Code. 11336198
Change view
Click to view available options
Quantity:
100 g
1 kg
Unit Size:
100g
1kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11336198 1 kg 1kg
11326198 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336198 Supplier Thermo Scientific Alfa Aesar Supplier No. 011181.A1
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 1314-36-9 | O3Y2 | 225.81 g/mol

Yttriium(III) oxide is used to make phosphors for color TV tubes, yttrium iron garnets for use as microwave filters, high temperature superconductors, as a mineralizing agent for the formation of spinel and as a stabilizer for high-temperature service materials. It is presurcor for important inorganic compounds. It is a potential solid-state laser material which allows the efficient operation both in continuous operation and in pulsed regimes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 1314-36-9
Molecular Formula O3Y2
Molecular Weight (g/mol) 225.81
MDL Number MFCD00011473
InChI Key RUDFQVOCFDJEEF-UHFFFAOYSA-N
IUPAC Name diyttrium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Y+3].[Y+3]

Spezifikation

Melting Point 2410°C
Physical Form Powder
Quantity 1 kg
Assay Percent Range 99.99% (REO)
Solubility Information Insoluble in water. Soluble in alcohol and dilute acids
Formula Weight 225.81
Density 5.01
Chemical Name or Material Yttrium(III) oxide

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.