missing translation for 'onlineSavingsMsg'
Learn More

Yttrium(III) perchlorate, 50% w/w aq. soln., Reagent Grade, Thermo Scientific™

Product Code. 11450311
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11450311

Brand: Thermo Scientific Alfa Aesar 040580.NB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14017-56-2
MDL Number MFCD00150430
Synonym yttrium perchlorate, acmc-20aj91, yttrium perchlorate 6h2o, yttrium iii perchlorate solution, perchloric acid yttrium iii salt, yttrium 3+ triperchlorate

Specifications

Color Colorless
Physical Form Liquid
Quantity 5 g
UN Number UN3211
Formula Weight 495.34 (387.25 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Chemical Name or Material Yttrium(III) perchlorate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.