missing translation for 'onlineSavingsMsg'
Learn More

Yttrium pieces, 99.9% (REO)

Product Code. 11346597
Change view
Click to view available options
Quantity:
10 g
50 g
Packaging:
Glass bottle; Ampouled under argon
Under Argon
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11346597 10 g Under Argon 10g
11336597 50 g Glass bottle; Ampouled under argon 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11346597 Supplier Thermo Scientific Alfa Aesar Supplier No. 000615.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7440-65-5 | Y | 88.91 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-65-5
Molecular Formula Y
Molecular Weight (g/mol) 88.91
MDL Number MFCD00011468
InChI Key VWQVUPCCIRVNHF-UHFFFAOYSA-N
Synonym yttrium, elemental, yttrium-89, ion, ytrio, hydride, unii-58784xqc3y, yttrium, metal and compounds, ingot, foil, element
PubChem CID 23993
ChEBI CHEBI:33331
IUPAC Name yttrium
SMILES [Y]

Specifications

Appearance Gray pieces
Density 4.47g/cm3
Quantity 10 g
Odor Odorless
Packaging Under Argon
Assay Percent Range 99.90%
Form Pieces
Chemical Name or Material Yttrium, Rare Earth Oxides Content

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.