missing translation for 'onlineSavingsMsg'
Learn More

(Z)-11-(3-(Dimethylamino)propylidene)-6,11-dihydrodibenzo[b,e]oxepine-2-carbaldehyde Hydrochloride, TRC

Product Code. 30012371
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30012371 2.5 mg 2.5mg
30004447 25 mg 25mg
30011767 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30012371 Supplier Toronto Research Chemicals Supplier No. TRCD4714102.5MG
 Restricted Item
Estimated Shipment: 26-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C20 H21 N O2 . Cl H
Molecular Weight (g/mol) 343.8472
Synonym Olopatadine Carbaldehyde Hydrochloride, Olopatadine Related Compound, Olopatadine Carbaldehyde (Z)-Isomer;
IUPAC Name (11Z)-11-[3-(dimethylamino)propylidene]-6H-benzo[c][1]benzoxepine-2-carbaldehyde;hydrochloride
SMILES Cl.CN(C)CC\C=C/1\c2ccccc2COc3ccc(C=O)cc13

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C20H21NO2.ClH/c1-21(2)11-5-8-18-17-7-4-3-6-16(17)14-23-20-10-9-15(13-22)12-19(18)20;/h3-4,6-10,12-13H,5,11,14H2,1-2H3;1H/b18-8-;
Formula Weight 343.1339
Percent Purity >95%
Chemical Name or Material (Z)-11-(3-(Dimethylamino)propylidene)-6,11-dihydrodibenzo[b,e]oxepine-2-carbaldehyde Hydrochloride
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.