missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies (Z)-4-Hydroxytamoxifen
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
Description
- (Z)-4-Hydroxytamoxifen is a cell-permeable selective estrogen receptor modulator (SERM) that binds to estrogen receptors (ER) and estrogen-related receptors (ERR) with both estrogenic and anti-estrogenic effects (Poulin et al.)
- Suppresses tumor growth in breast cancer cells (Freiss et al.; Osborne et al.)
Specifications
Specifications
| CAS | 68047-06-3 |
| Synonym | trans-4-Hydroxytamoxifen; 4-OH-TAM; ICI 79280 |
| Quantity | 5 mg |
| Molecular Formula | C26H29NO2 |
| Purity | ≥98% |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction