missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone

Product Code. 11434906 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11434906 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11434906 Supplier MP Biomedicals™ Supplier No. 03FK01001
  • ONLINE EXCLUSIVE PRICE
    543.00 €
    514.50 € / 1mg
    Save 28.50 €
    5% Off
 Restricted Item
Estimated Shipment: 29-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-FMK is an irreversible and cell permeable inhibitor of CPP32/Apopain.

A member of the CIE/CED-3 family of cysteine proteases.

Specifications

Color White
Content And Storage Store in a cool, dry place. Dessicate.
Quantity 1 mg
Product Type Z-Asp(OMe)-Glu(OMe)-Val-Asp(OMe)-Fluoromethylketone
Molecular Weight (g/mol) 668
Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.