missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Z-D-Phenylalaninol, TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Z-D-Phenylalaninol |
| CAS | 58917-85-4 |
| Molecular Formula | C17H19NO3 |
| Quantity | 1 g |
| Synonym | (R)-[1-(Hydroxymethyl)-2-phenylethyl]-carbamic Acid Phenylmethyl Ester, [(1R)-1-(Hydroxymethyl)-2-phenylethyl]-carbamic Acid Phenylmethyl Ester, R)-Z-Phenylalaninol, N-Benzyloxycarbonyl-D-phenylalaninol |
| InChI Formula | InChI=1S/C17H19NO3/c19-12-16(11-14-7-3-1-4-8-14)18-17(20)21-13-15-9-5-2-6-10-15/h1-10,16,19H,11-13H2,(H,18,20)/t16-/m1/s1 |
| SMILES | O=C(OCC1=CC=CC=C1)N[C@@H](CO)CC2=CC=CC=C2 |
| IUPAC Name | benzyl (R)-(1-hydroxy-3-phenylpropan-2-yl)carbamate |
| Molecular Weight (g/mol) | 285.34 |
| Formula Weight | 285.14 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction