missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Z-epsilon-Acp-OH, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | N-Benzyloxycarbonyl-6-aminocaproic Acid |
| CAS | 1947-00-8 |
| Molecular Formula | C14 H19 N O4 |
| Quantity | 5 g |
| Synonym | 6-[[(Phenylmethoxy)carbonyl]amino]-hexanoic Acid, 6-(N-Benzyloxycarbonylamino)hexanoic Acid, N-(Benzyloxycarbonyl)-6-aminohexanoic Acid, NSC 92812, VIB 19, N-Benzyloxycarbonyl-6-aminocaproic Acid |
| InChI Formula | InChI=1S/C14H19NO4/c16-13(17)9-5-2-6-10-15-14(18)19-11-12-7-3-1-4-8-12/h1,3-4,7-8H,2,5-6,9-11H2,(H,15,18)(H,16,17) |
| SMILES | OC(=O)CCCCCNC(=O)OCc1ccccc1 |
| IUPAC Name | 6-(phenylmethoxycarbonylamino)hexanoic acid |
| Molecular Weight (g/mol) | 265.3 |
| Formula Weight | 265.1314 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction